missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATH2/NEUROD6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-29865-0.025MG
This item is not returnable.
View return policy
Description
MATH2/NEUROD6 Polyclonal specifically detects MATH2/NEUROD6 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| MATH2/NEUROD6 | |
| Polyclonal | |
| Western Blot 1:200-1:1000, Immunohistochemistry 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin | |
| Q96NK8 | |
| NEUROD6 | |
| Synthetic peptide. | |
| 0.025 mg | |
| Primary | |
| Recognizes NeuroD 6 (MATH2, NEX1), a Helix-loop-helix class of transcription factor. | |
| Store at -70°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 50% saturated ammonium sulfate and PBS with No Preservative | |
| Atoh2, BHLHA2, bHLHa2MATH2, Class A basic helix-loop-helix protein 2, Math-2, NeuroD6, neurogenic differentiation 6, neurogenic differentiation factor 6, NEX1M, Protein atonal homolog 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 63974 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction