missing translation for 'onlineSavingsMsg'
Learn More

MAT2A Antibody (SR1837), Novus Biologicals™

Rabbit Monoclonal Antibody

349.00 € - 449.00 €

Specifications

Antigen MAT2A
Clone SR1837
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Applications Western Blot, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18671214
View Documents
Novus Biologicals
NBP3-22383-100ul
100 μg
449.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18641845
View Documents
Novus Biologicals
NBP3-22383-50ul
50 μg
349.00 €
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MAT2A Monoclonal antibody specifically detects MAT2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

MAT2A
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Monoclonal
Purified
RUO
Human, Mouse, Rat
AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2
A synthesized peptide derived from human MAT2A (Uniprot #: P31153)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR1837
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Signal Transduction
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
4144
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.