missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAT2A Antibody (SR1837), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22383-50ul
This item is not returnable.
View return policy
Description
MAT2A Monoclonal antibody specifically detects MAT2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| MAT2A | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR1837 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2 | |
| A synthesized peptide derived from human MAT2A (Uniprot #: P31153) | |
| 50 μg | |
| Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Signal Transduction | |
| 4144 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction