Learn More
Maprotiline EP Impurity B (as Hydrochloride), Mikromol™
Description
Maprotiline EP Impurity B (as Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | 3-(9,10-Ethanoanthracen-9(10H)-yl)-N-[3-(9,10-ethanoanthracen-9(10H)-yl)propyl]-N-methylpropan-1-amine Hydrochloride |
| CAS | 2724727-05-1 |
| Analyte or Component Names | 3-(9,10-Ethanoanthracen-9(10H)-yl)-N-[3-(9,10-ethanoanthracen-9(10H)-yl)propyl]-N-methylpropan-1-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C39 H41 N . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Maprotiline Hydrochloride Imp. B (EP), Maprotiline Imp. B (EP), 3-(9,10-Ethanoanthracen-9(10H)-yl)-N-[3-(9,10-ethanoanthracen-9(10H)-yl)propyl]-N-methylpropan-1-amine Hydrochloride, Maprotiline Hydrochloride Imp. B (EP) as Hydrochloride, Maprotiline Hydrochloride Impurity B as Hydrochloride, Maprotiline Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C19H21ClN2O2.2ClH/c20-17-8-6-16(7-9-17)19(15-4-2-1-3-5-15)22-12-10-21(11-13-22)14-18(23)24;;/h1-9,19H,10-14H2,(H,23,24);2*1H |
| SMILES | Cl.CN(CCCC12CCC(c3ccccc13)c4ccccc24)CCCC56CCC(c7ccccc57)c8ccccc68 |
| Molecular Weight (g/mol) | 560.21 |
| Show More |