Learn More
Levomepromazine Sulfone N-Oxide (Levomepromazine N,S,S-Trioxide), Mikromol™
Description
Levomepromazine Sulfone N-Oxide (Levomepromazine N,S,S-Trioxide)
Specifications
Specifications
| Chemical Name or Material | Levomepromazine Sulfone N-Oxide |
| CAS | 2724689-26-1 |
| Analyte or Component Names | Levomepromazine Sulfone N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H24 N2 O4 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Levomepromazine Sulfone N-Oxide |
| InChI Formula | InChI=1S/C19H24N2O4S/c1-14(13-21(2,3)22)12-20-16-7-5-6-8-18(16)26(23,24)19-10-9-15(25-4)11-17(19)20/h5-11,14H,12-13H2,1-4H3/t14-/m0/s1 |
| SMILES | COc1ccc2c(c1)N(C[C@H](C)C[N+](C)(C)[O-])c3ccccc3S2(=O)=O |
| IUPAC Name | (2S)-3-(2-methoxy-5,5-dioxophenothiazin-10-yl)-N,N,2-trimethylpropan-1-amine oxide |
| Show More |