missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leptin R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
213.00 €
Specifications
| Antigen | Leptin R |
|---|---|
| Concentration | 10 mg/mL |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
Leptin R Polyclonal specifically detects Leptin R in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| Leptin R | |
| Western Blot 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| P48357 | |
| 3953 | |
| Synthetic peptide corresponding to residues K(577) E I Q W K T H E V F D A K S K S A(594) of the extracellular domain of rat OB-R. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 10 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS with 0.05% Sodium Azide | |
| CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
| LEPR | |
| IgG | |
| Ammonium sulfate precipitation | |
| Leptin Receptor |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title