missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leptin R Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB120-5593-0.025ml
This item is not returnable.
View return policy
Description
Leptin R Polyclonal specifically detects Leptin R in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Leptin R | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| CD295 antigen, DB, DKFZp686B1731, huB219, LEP-R, leptin receptor, OB receptor, OB-R, OBRCD295 | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Leptin Receptor | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| 10 mg/mL | |
| Western Blot 1:2000 | |
| P48357 | |
| LEPR | |
| Synthetic peptide corresponding to residues K(577) E I Q W K T H E V F D A K S K S A(594) of the extracellular domain of rat OB-R. | |
| 0.025 mL | |
| Signal Transduction | |
| 3953 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction