missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin A + C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
226.00 € - 372.00 €
Specifications
| Antigen | Lamin A + C |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Flow (Intracellular) 1:500 - 1:1000 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18067063
|
Novus Biologicals
NB100-56649SS |
0.05 mL |
226.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18700504
|
Novus Biologicals
NB100-56649 |
0.2 mL |
372.00 €
0.20mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lamin A + C Polyclonal specifically detects Lamin A + C in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).Specifications
| Lamin A + C | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P02545 | |
| 4000 | |
| A synthetic peptide corresponding to amino acids 249-265 CRAQHEDQVEQYKKELEK of Human Lamin A/C was used immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Flow (Intracellular) 1:500 - 1:1000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Whole antisera with PBS and 50% Glycerol with 0.05% Sodium Azide | |
| CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32 | |
| LMNA | |
| IgG | |
| Unpurified | |
| Because of immunogen's 100% homology with Isoforms 1, 4, 5, 6, ADelta10 and C of Prelamin-A/C, this antibody is expected to detect multiple bands in WB. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title