missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lamin A + C Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB100-56649
This item is not returnable.
View return policy
Description
Lamin A + C Polyclonal specifically detects Lamin A + C in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Flow (Intracellular).
Specifications
| Lamin A + C | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1:100, Flow (Intracellular) 1:500 - 1:1000 | |
| P02545 | |
| LMNA | |
| A synthetic peptide corresponding to amino acids 249-265 CRAQHEDQVEQYKKELEK of Human Lamin A/C was used immunogen. | |
| 0.2 mL | |
| Primary | |
| Because of immunogen's 100% homology with Isoforms 1, 4, 5, 6, ADelta10 and C of Prelamin-A/C, this antibody is expected to detect multiple bands in WB. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence, Flow Cytometry | |
| Unconjugated | |
| Whole antisera with PBS and 50% Glycerol with 0.05% Sodium Azide | |
| CDCD1, CDDC, CMD1A, CMT2B1, dilated 1A (autosomal dominant), EMD2, FPLD, HGPSFPL, lamin A/C, lamin A/C-like 1, lamin-A/C, LDP1, LFP, LGMD1B, limb girdle muscular dystrophy 1B (autosomal dominant), LMN1IDC, LMNC, LMNL1, prelamin-A/C, PRO1,70 kDa lamin, progeria 1 (Hutchinson-Gilford type), renal carcinoma antigen NY-REN-32 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| 4000 | |
| Human | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction