Learn More
Lactam of (S)-Carboxymethyl-L-cysteine, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Lactam of (S)-Carboxymethyl-L-cysteine
Specifications
Specifications
| Chemical Name or Material | Lactam of (S)-Carboxymethyl-L-cysteine |
| CAS | 62305-89-9 |
| Analyte or Component Names | Lactam of (S)-Carboxymethyl-L-cysteine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H7 N O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | Lactam of (S)-Carboxymethyl-L-cysteine, 3-Thiomorpholinecarboxylic acid, 5-oxo-, (3R)-, 3-Thiomorpholinecarboxylic acid, 5-oxo-, (R)-, (3R)-3-Carboxy-5-oxothiomorpholine |
| InChI Formula | InChI=1S/C5H7NO3S/c7-4-2-10-1-3(6-4)5(8)9/h3H,1-2H2,(H,6,7)(H,8,9)/t3-/m0/s1 |
| SMILES | OC(=O)[C@@H]1CSCC(=O)N1 |
| IUPAC Name | (3R)-5-oxothiomorpholine-3-carboxylic acid |
| Show More |