missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Calretinin Rabbit Monoclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ Calretinin, Rabbit Monoclonal Antibody.

177.70 € - 741.49 €

Specifications

Antigen Calretinin
Clone SP13
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Product Code Brand Quantity Form Price Quantity & Availability  
Product Code Brand Quantity Form Price Quantity & Availability  
12693727
View Documents
Epredia™
RM-9113-S0
100 μL Concentrated
ONLINE EXCLUSIVE PRICE
191.00 €
177.70 € / 100µL
Save 13.30 €
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12603737
View Documents
Epredia™
RM-9113-S1
500 μL Concentrated
ONLINE EXCLUSIVE PRICE
522.00 €
473.13 € / 500µL
Save 48.87 €
9% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12673727
View Documents
Epredia™
RM-9113-S
1 mL Concentrated
ONLINE EXCLUSIVE PRICE
812.00 €
741.49 € / 1mL
Save 70.51 €
8% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
12683727
View Documents
Epredia™
RM-9113-R7
7 mL Prediluted This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
12663727
View Documents
Epredia™
RM-9113-RQ
12 mL Ready-to-Use
ONLINE EXCLUSIVE PRICE
697.00 €
649.33 € / 12mL
Save 47.67 €
6% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.

Host Species: Rabbit

Clone: SP13

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length mouse calretinin protein

Molecular Weight: 31.5 kDa

Positive Control: Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

Calretinin
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Recombinant full-length mouse calretinin protein
SP13
Monoclonal
Rabbit
Oncogenesis
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.