missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ Calretinin Rabbit Monoclonal Antibody

Ensure accurate and reproducible results in immunohistochemistry experiments with Epredia™ Calretinin, Rabbit Monoclonal Antibody.

Brand:  Epredia™ RM-9113-S

 View more versions of this product

Product Code. 12673727

  • ONLINE EXCLUSIVE PRICE
    812.00 €
    741.49 € / 1mL
    Save 70.51 €
    8% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Calretinin is an intracellular calcium-binding protein belonging to the troponin C superfamily characterized by a structural motif described as the EF-hand domain. The immunohistochemical detection of calretinin in developing cerebellum is restricted to the later stages indicated by weak staining from week 21 of gestation, in Purkinje and basket cells and in neurons of the dentate nucleus. The intensity of staining increases as the cerebellum matures. In tumors, calretinin has been detected in mesotheliomas and some pulmonary adenocarcinomas.

Host Species: Rabbit

Clone: SP13

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Not determined

Immunogen: Recombinant full-length mouse calretinin protein

Molecular Weight: 31.5 kDa

Positive Control: Mesothelioma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calretinin
Monoclonal
Unconjugated
Recombinant full-length mouse calretinin protein
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
SP13
Rabbit
1 mL
Oncogenesis
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.