Learn More
L-Phenylalanine-d7, TRC
Product Code. 30179232
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179232 5 mg 5mg
30179117 25 mg 25mg
30179075 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30179232 Supplier Toronto Research Chemicals Supplier No. TRCP3194165MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 69113-60-6
Molecular Formula C9H4D7NO2
Molecular Weight (g/mol) 172.23
Synonym (2S)-2-Amino-3-phenylpropanoic Acid-d7, (S)-(-)-Phenylalanine-d7, (S)-α-Amino-β-phenylpropionic Acid-d7, (S)-α-Aminohydrocinnamic Acid-d7, (S)-α-Amino-benzenepropanoic Acid-d7;
SMILES N[C@H](C(O)=O)C([2H])([2H])C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1/i1D,2D,3D,4D,5D,6D2
Formula Weight 172.1229
Percent Purity >95%
Chemical Name or Material L-Phenylalanine-d7
Grade HPLC