CAS RN 69113-60-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Phenylalanine-d7, TRC
CAS: 69113-60-6 Molecular Formula: C9H4D7NO2 Molecular Weight (g/mol): 172.23 Synonym: (2S)-2-Amino-3-phenylpropanoic Acid-d7,(S)-(-)-Phenylalanine-d7,(S)-α-Amino-β-phenylpropionic Acid-d7,(S)-α-Aminohydrocinnamic Acid-d7,(S)-α-Amino-benzenepropanoic Acid-d7; SMILES: N[C@H](C(O)=O)C([2H])([2H])C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H]