Learn More
L-Alanyl-L-alanyl-L-alanine, TRC
Product Code. 30179225
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179225 50 mg 50mg
30179074 100 mg 100mg
30179127 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30179225 Supplier Toronto Research Chemicals Supplier No. TRCA57639350MG
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5874-90-8
Molecular Formula C9H17N3O4
Molecular Weight (g/mol) 231.25
Synonym Alanine, N-(N-L-alanyl-L-alanyl)-, L- (8CI), Alanine, N-(N-alanylalanyl)- (7CI), L-Alanine, N-(N-L-alanyl-L-alanyl)-, Alanylalanylalanine, L-Ala-L-ala-L-ala, L-Alanine trimer, L-Alanine-L-alanine-L-alanine, Tri-L-alanine, Trialanine
IUPAC Name (2S)-2-[[(2S)-2-[[(2S)-2-aminopropanoyl]amino]propanoyl]amino]propanoic acid
SMILES C[C@H](N)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)O
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C9H17N3O4/c1-4(10)7(13)11-5(2)8(14)12-6(3)9(15)16/h4-6H,10H2,1-3H3,(H,11,13)(H,12,14)(H,15,16)/t4-,5-,6-/m0/s1
Formula Weight 231.1219
Chemical Name or Material L-Alanyl-L-alanyl-L-alanine