CAS RN 5874-90-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
L-Alanyl-L-alanyl-L-alanine, TRC
CAS: 5874-90-8 Molecular Formula: C9H17N3O4 Molecular Weight (g/mol): 231.25 Synonym: Alanine, N-(N-L-alanyl-L-alanyl)-, L- (8CI),Alanine, N-(N-alanylalanyl)- (7CI),L-Alanine, N-(N-L-alanyl-L-alanyl)-,Alanylalanylalanine,L-Ala-L-ala-L-ala,L-Alanine trimer,L-Alanine-L-alanine-L-alanine,Tri-L-alanine,Trialanine IUPAC Name: (2S)-2-[[(2S)-2-[[(2S)-2-aminopropanoyl]amino]propanoyl]amino]propanoic acid SMILES: C[C@H](N)C(=O)N[C@@H](C)C(=O)N[C@@H](C)C(=O)O