Learn More
KN-92 phosphate, MedChemExpress
Product Code. 16450246
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16450246 2 mg 2mg
16432687 50 mg 50mg
16409167 5 mg 5mg
16406598 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16450246 Supplier Medchem Express Supplier No. HY15517A
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress KN-92 phosphate is an inactive derivative of KN-93, without CaM kinase inhibitory activity. KN-92 phosphate is intended to be used as a control compound in studies designed to elucidate the antagonist activities of KN-93. KN-93 is a cell-permeable, reversible and competitive CaMKII inhibitor.
KN-92 phosphate is an inactive derivative of KN-93, without CaM kinase inhibitory activity. KN-92 phosphate is intended to be used as a control compound in studies designed to elucidate the antagonist activities of KN-93. KN-93 is a cell-permeable, reversible and competitive CaMKII inhibitor.
Specifications
Chemical Name or Material KN-92 phosphate
CAS 1135280-28-2
Color White
Purity Grade Notes Research
Molecular Formula C24H28ClN2O7PS
Quantity 2 mg
Solubility Information DMSO : 100 mg/mL (180.19 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic) (insoluble)
SMILES O=S(C1=CC=C(OC)C=C1)(NC2=CC=CC=C2CN(C/C=C/C3=CC=C(Cl)C=C3)C)=O.O=P(O)(O)O
Molecular Weight (g/mol) 554.98
Formula Weight 554.98
Percent Purity 99.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease