missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KML 29
GREENER_CHOICE

Product Code. 30002032
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002032 10 mg 10mg
30002040 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002032 Supplier Tocris Bioscience™ Supplier No. 4872/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent and selective MAGL inhibitor

Highly selective and potent monoacylglycerol lipase (MAGL) inhibitor. Exhibits potent inhibition of human, mouse and rat MAGL (IC50 values are 5.9, 15 and 43 nM, respectively). Exhibits no detectable inhibition of FAAH (IC50 > 50000 nM). Potently and selectively blocks hydrolysis of 2-arachidonoylglycerol (2-AG) in mice (IC50 = 2.5 nM and >50 μM for 2-AG and AEA respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1380424-42-9
Molecular Formula C24H21F6NO7

Specifications

Quantity 10 mg
Formula Weight 549.42
Molecular Weight (g/mol) 549.422
Chemical Name or Material KML 29
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.