missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Kjeldahl Tablets Titanium

Product Code. 18845423 Shop All Buchi Products
Change view
Click to view available options
Quantity:
1000/Cs.
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18845423 1000/Cs. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18845423 Supplier BUCHI Supplier No. 11072627
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Kjeldahl Tablets Titanium

  • Digestion aids for nitrogen determination.
  • Environmentally friendly: free of toxic mercury and selenium
  • Different tablets to fit individual needs
  • For digestion
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material K2SO4, CuSO4 - 5H2O, TiO2
Color Blue
Quantity 1000/Cs.
CAS 7778-80-5, 7758-99-8, 13463-67-7
Packaging Plastic Container
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.