missing translation for 'onlineSavingsMsg'
Learn More

Kira8, MedChemExpress

Product Code. 16457567
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16457567

Brand: Medchem Express HY114368

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Kira8 (AMG-18) is a mono-selective IRE1α inhibitor that allosterically attenuates IRE1α RNase activity with an IC50 of 5.9 nM.

Kira8 (AMG-18) is a mono-selective IRE1α inhibitor that allosterically attenuates IRE1α RNase activity with an IC50 of 5.9 nM.

Specifications

Chemical Name or Material Kira8
CAS 1630086-20-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C31H29ClN6O3S
Quantity 5 mg
Synonym AMG-18
Solubility Information DMSO : 65 mg/mL (108.13 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC=C1Cl)(NC2=C3C=CC(C)=C(OC4=NC=CC=C4C5=NC(N[C@@H]6CNCCC6)=NC=C5)C3=CC=C2)=O
Molecular Weight (g/mol) 601.12
Formula Weight 601.12
Percent Purity 98.02%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.