Learn More
Ketoprofen EP Impurity G, Mikromol™
Product Code. 30458009
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30458009 4 x 25 mg 100mg
17971426 25 mg 25mg
17983786 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30458009 Supplier Mikromol Supplier No. MM0001.2000254AMP
Please to purchase this item. Need a web account? Register with us today!
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Ketoprofen EP Impurity G, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-[(1RS)-1-Cyanoethyl]benzoic Acid
CAS 5537-71-3
Analyte or Component Names 3-[(1RS)-1-Cyanoethyl]benzoic Acid
Shipping Condition Room Temperature
Molecular Formula C10 H9 N O2
Certifications/Compliance ISO 17025
Synonym 3-[(1RS)-1-Cyanoethyl]benzoic Acid, Ketoprofen Imp. G (EP), Ketoprofen Impurity G
InChI Formula InChI=1S/C10H9NO2/c1-7(6-11)8-3-2-4-9(5-8)10(12)13/h2-5,7H,1H3,(H,12,13)
Solution Type Neat
SMILES CC(C#N)c1cccc(c1)C(=O)O
IUPAC Name 3-(1-cyanoethyl)benzoic acid
Molecular Weight (g/mol) 175.18
Quantity 4 x 25 mg
Formula Weight 175.0633 g/mol