Learn More
Ketoconazole EP Impurity D, Mikromol™
Description
Ketoconazole EP Impurity D
Specifications
Specifications
| Chemical Name or Material | Deacetyl Ketoconazole |
| CAS | 67914-61-8 |
| Analyte or Component Names | Deacetyl Ketoconazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H26 Cl2 N4 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-[4-[[(2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, Ketoconazole Imp. D (EP), 1-[4-[[(2RS,4SR)-2-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine, Ketoconazole Impurity D |
| InChI Formula | InChI=1S/C24H26Cl2N4O3/c25-18-1-6-22(23(26)13-18)24(16-29-10-7-28-17-29)32-15-21(33-24)14-31-20-4-2-19(3-5-20)30-11-8-27-9-12-30/h1-7,10,13,17,21,27H,8-9,11-12,14-16H2/t21-,24-/m0/s1 |
| SMILES | Clc1ccc(c(Cl)c1)[C@@]2(Cn3ccnc3)OC[C@H](COc4ccc(cc4)N5CCNCC5)O2 |
| IUPAC Name | 1-[4-[[(2R,4S)-2-(2,4-dichlorophenyl)-2-(imidazol-1-ylmethyl)-1,3-dioxolan-4-yl]methoxy]phenyl]piperazine |
| Show More |