Learn More
IUPAC pH Standard pH 6.865, Hach Lange™
Product Code. 10503923
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10503923 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10503923 Supplier Hach Lange Supplier No. S11M003
Please to purchase this item. Need a web account? Register with us today!
Edge
With an accuracy of 10.012 ± 0.010 and a pH of 6.865, this pH standard buffer is traceable to IUPAC pH scale and NIST.
  • pH Standard, IUPAC, pH10.012 at 25 °C, Certified, 500 mL (Radiometer Analytical) 
  • 10.012 ± 0.010 pH at 25°C concentration
  • Traceable to IUPAC pH scale and NIST
  • Shelf life of 48 months from production date
  • Each bottle (500 mL) is supplied with a COFRAC Verification Certificate and Certificate of Conformity and Traceability drawn up according to ISO31
  • Direct traceability to primary standards and the reference method
  • The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference materials
TRUSTED_SUSTAINABILITY
Specifications
pH 6.865
Physical Form Liquid
Quantity 500 mL
Traceability to NIST Yes