Learn More
Isoconazole Nitrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Isoconazole Nitrate
Specifications
Specifications
| Chemical Name or Material | Isoconazole Nitrate |
| CAS | 24168-96-5 |
| Analyte or Component Names | Isoconazole Nitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H14 Cl4 N2 O . H N O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Isoconazole Nitrate, 1-[(2RS)-2-[(2,6-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate, Miconazole Nitrate Imp. D (EP) as Nitrate, Miconazole Impurity D as Nitrate, Miconazole Nitrate Impurity D as Nitrate, Miconazole Impurity D as Nitrate |
| InChI Formula | InChI=1S/C18H14Cl4N2O.HNO3/c19-12-4-5-13(17(22)8-12)18(9-24-7-6-23-11-24)25-10-14-15(20)2-1-3-16(14)21;2-1(3)4/h1-8,11,18H,9-10H2;(H,2,3,4) |
| SMILES | O[N+](=O)[O-].Clc1ccc(C(Cn2ccnc2)OCc3c(Cl)cccc3Cl)c(Cl)c1 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-[(2,6-dichlorophenyl)methoxy]ethyl]imidazole;nitric acid |
| Show More |