Learn More
Isoconazole EP Impurity B (as Nitrate), Mikromol™
Description
Isoconazole EP Impurity B (as Nitrate), Mikromol
Specifications
Specifications
| Chemical Name or Material | (2RS)-2-[(2,6-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine Nitrate |
| CAS | 2726492-80-2 |
| Analyte or Component Names | (2RS)-2-[(2,6-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine Nitrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H13 Cl4 N O . H N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Isoconazole Nitrate Imp. B (EP), (2RS)-2-[(2,6-Dichlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethanamine Nitrate, Isoconazole Nitrate Imp. B (EP) as Nitrate, Isoconazole Nitrate Impurity B as Nitrate, Isoconazole Impurity B as Nitrate |
| InChI Formula | InChI=1S/C19H21ClN2.ClH/c1-21(2)12-5-13-22-18-7-4-3-6-15(18)8-9-16-10-11-17(20)14-19(16)22;/h3-4,6-11,14H,5,12-13H2,1-2H3;1H |
| Solution Type | Neat |
| SMILES | NCC(OCc1c(Cl)cccc1Cl)c2ccc(Cl)cc2Cl.O[N+](=O)[O-] |
| Show More |