Learn More
Irbesartan-d4, MedChemExpress
Description
Irbesartan D4 is the deuterium labeled Irbesartan, which is a highly potent and specific angiotensin II type 1 (AT1) receptor antagonist.
Specifications
Specifications
| Chemical Name or Material | Irbesartan-d4 |
| CAS | 1216883-23-6 |
| Purity Grade Notes | Research |
| Physical Form | Solid |
| Molecular Formula | C25H24D4N6O |
| Color | White |
| Synonym | SR-47436-d4 BMS-186295-d4 |
| SMILES | O=C1N(CC2=C([2H])C([2H])=C(C3=C(C4=NN=NN4)C=CC=C3)C([2H])=C2[2H])C(CCCC)=NC15CCCC5 |
| Industry Type | Neuroscience-Neuromodulation |
| Molecular Weight (g/mol) | 432.55 |
| Show More |