Learn More
Memmert™ Iq/Oq Document W/Device-Specific Works Test Data (Temp., Humidity And Light Value), For Ich750Eco
Shop All Memmert ProductsDescription
Includes
Temperature distribution survey for one free-selectable temperature, humidity and light value. 27 measuring points to DIN 12880:2007-05, PQ check list as support for validation by customer (models HPP with light and ICH L)
- IQ/OQ document with works test data for chamber
- 27 measuring points to DIN 12880:2007-05
- PQ check list as support for validation by customer (models HPP with light and ICH L)
Specifications
Specifications
| Product Type | IQ/OQ Document with Works Test Data for Chamber |