Learn More
Memmert™ IQ Document with Test Data for SF160
Product Code. 17508135 Shop All Memmert Products
Change view
Click to view available options
:
For Use With: SF160
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
17508135 For Use With: SF160
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17508135 Supplier Memmert™ Supplier No. SF160D00124
Please to purchase this item. Need a web account? Register with us today!
IQ document with device-specific works test data for validation by customer.
  • OQ/PQ check list as support for validation by customer
Specifications
For Use With (Equipment) SF160 Sterilizer
Product Type IQ Document
Includes Test Data