Learn More
Ioxitalamic acid, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Ioxitalamic Acid |
| CAS | 28179-44-4 |
| Analyte or Component Names | Ioxitalamic Acid |
| Shipping Condition | +20°C |
| Molecular Formula | C12 H11 I3 N2 O5 |
| Certifications/Compliance | ISO/IEC 17025 |
| Synonym | 3-(Acetylamino)-5-[[(2-hydroxyethyl)amino]carbonyl]-2,4,6-triiodobenzoic Acid, 3-(Acetylamino)-5-[[(2-hydroxyethyl)amino]carbonyl]-2,4,6-triiodo-benzoic acid, Isophthalamic acid, 5-acetamido-N-(2-hydroxyethyl)-2,4,6-triiodo- (8CI), AG 58107, Ioxitalamate, Ioxitalamic acid, Ioxithalaminic acid, 3-Acetamido-2,4,6-triiodo-(N-beta-hydroxyethyl)isophthalic acid monoamide |
| InChI Formula | InChI=1S/C12H11I3N2O5/c1-4(19)17-10-8(14)5(11(20)16-2-3-18)7(13)6(9(10)15)12(21)22/h18H,2-3H2,1H3,(H,16,20)(H,17,19)(H,21,22) |
| SMILES | CC(=O)Nc1c(I)c(C(=O)O)c(I)c(C(=O)NCCO)c1I |
| IUPAC Name | 3-acetamido-5-(2-hydroxyethylcarbamoyl)-2,4,6-triiodobenzoic acid |
| Show More |