Learn More
IL-4 Rat anti-Human, FITC, Clone: MP4-25D2, BD
Rat Monoclonal Antibody
93.09 € - 415.00 €
Specifications
| Antigen | IL-4 |
|---|---|
| Clone | MP4-25D2 |
| Concentration | 0.5mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15801599
|
BD Biosciences
562047 |
25 μg |
94.20 € 93.09 € / Each Save 1.11 € 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15857728
|
BD Biosciences
554484 |
0.1 mg |
415.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MP4-25D2 antibody reacts with human interleukin-4 (IL-4). The immunogen used to generate the MP4-25D2 hybridoma was purified recombinant human IL-4. This is a neutralizing antibody. The MP4-25D2 antibody has been reported to cross react with IL-4 from rhesus monkeys. The use of the MP4-25D2 antibody for epitope mapping of human IL-4 has been described.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| IL-4 | |
| 0.5mg/mL | |
| Monoclonal | |
| Rat | |
| Human | |
| Purified Recombinant Human IL-4 | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| MP4-25D2 | |
| Flow Cytometry | |
| FITC | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.