Learn More
IL-4 Rat anti-Human, FITC, Clone: MP4-25D2, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554484
Description
The MP4-25D2 antibody reacts with human interleukin-4 (IL-4). The immunogen used to generate the MP4-25D2 hybridoma was purified recombinant human IL-4. This is a neutralizing antibody. The MP4-25D2 antibody has been reported to cross react with IL-4 from rhesus monkeys. The use of the MP4-25D2 antibody for epitope mapping of human IL-4 has been described.
Host Species: Rat
Clone: MP4-25D2
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified Recombinant Human IL-4
Intracellular Staining
Specifications
| IL-4 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Purified Recombinant Human IL-4 | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| MP4-25D2 | |
| FITC | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.