Learn More
Thermo Scientific™ iConnect™ Flame Ionization Detector (FID)
Product Code. 31903476 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
TRACE 1300/1600 Series GC
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
31903476 TRACE 1300/1600 Series GC Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31903476 Supplier Thermo Scientific™ Supplier No. 19070001
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
iConnect Flame Ionization Detector (FID) enables highly sensitive detection of organic compounds with a wide linear dynamic range, fast acquisition speed, and flexible modular integration for efficient GC workflows.
  • Flame ionization detector (FID) module designed for highly sensitive detection of organic compounds
  • Linear dynamic range greater than 107(±10%) enables accurate quantification of analytes across a broad concentration range
  • High sensitivity with minimum detectable level (MDL) less than 1.2 pg C/s and sensitivity greater than 0.03 Coulombs/gC
  • Small cell volume and rapid acquisition rate (up to 600 Hz) support both standard and fast GC applications
  • Maximum operating temperature up to 450°C
  • Integrated electronic gas control (IEC) for precise regulation of air (0 to 500 mL/min), hydrogen (0 to 100 mL/min), and makeup gas (N2 or He, 0 to 50 mL/min)
  • Flameout detection with automatic re-ignition improves operational reliability and reduces downtime
  • Optimized for capillary columns and compatible with 1/8 in. and 1/16 in. packed columns
  • Modular iConnect design allows tool-free installation and replacement in minutes, enabling rapid system reconfiguration
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) TRACE 1300/1600 Series GC
Product Type FID Detector