missing translation for 'onlineSavingsMsg'
Learn More

ICA-121431, MedChemExpress

Product Code. 16457198
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16457198

Brand: Medchem Express HY16787

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ICA-121431 is a nanomolar potent and broad-spectrum voltage-gated sodium channel (Nav) blocker, shows equipotent selectivity for human Nav1.1 and Nav1.3 subtypes with IC50 values of 13 nM and 23 nM, respectively. ICA-121431 shows less potent inhibition of Nav1.2 (IC50=240 nM) and 1,000 fold selectivity against Nav1.4, Nav1.6, and the TTX-resistant human Nav1.5 and Nav1.8 channels (IC50s >10 μM).

ICA-121431 is a nanomolar potent and broad-spectrum voltage-gated sodium channel (Nav) blocker, shows equipotent selectivity for human Nav1.1 and Nav1.3 subtypes with IC50 values of 13 nM and 23 nM, respectively. ICA-121431 shows less potent inhibition of Nav1.2 (IC50=240 nM) and 1,000 fold selectivity against Nav1.4, Nav1.6, and the TTX-resistant human Nav1.5 and Nav1.8 channels (IC50s >10 μM).

Specifications

Chemical Name or Material ICA-121431
CAS 313254-51-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H19N3O3S2
Quantity 10 mg
Solubility Information DMSO : ≥ 44 mg/mL (97.88 mM)
SMILES O=C(NC1=CC=C(S(NC2=NC=CS2)(=O)=O)C=C1)C(C3=CC=CC=C3)C4=CC=CC=C4
Molecular Weight (g/mol) 449.55
Formula Weight 449.55
Percent Purity 98.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.