missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ibuprofen PEG Ester, TRC
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
Specifications
Specifications
| Chemical Name or Material | Ibuprofen PEG Ester |
| Analyte or Component Names | Ibuprofen PEG Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29H50O10 |
| Synonym | 23-hydroxy-3,6,9,12,15,18,21-heptaoxatricosyl 2-(4-isobutylphenyl)propanoate |
| InChI Formula | InChI=1S/C29H50O10/c1-25(2)24-27-4-6-28(7-5-27)26(3)29(31)39-23-22-38-21-20-37-19-18-36-17-16-35-15-14-34-13-12-33-11-10-32-9-8-30/h4-7,25-26,30H,8-24H2,1-3H3 |
| Solution Type | Neat |
| SMILES | O=C(C(C1=CC=C(CC(C)C)C=C1)C)OCCOCCOCCOCCOCCOCCOCCOCCO |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 558.71 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction