Learn More
Thermo Scientific™ HyPURITY™ C18 Columns, 3μm Particle Size
Product Code. 10359293 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
30 mm
125 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
4 mm
4.6 mm
Unit Size:
Each
Product Code. 10359293
Brand: Thermo Scientific™ 22103154030
Only null left
Add to basket
Add to basket
High quality, highly pure silica columns
  • Highly characterized, tested and validated C18 columns
Order Info
Other column dimensions are also available. Please call Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Column Format Analytical Column
Diameter (Metric) 4 mm
Length (Metric) 150 mm
Particle Size 3 μm
pH 1 to 9
Pore Size 190 Å
Surface Area 200 m²/g
USP Type L1
Stationary Phase C18
Packing Material Silica
Carbon Load 13%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase