Learn More
Hydroxychloroquine Sulfate EP Impurity C (as Dihydrochloride), Mikromol™
Description
Hydroxychloroquine Sulfate EP Impurity C (as Dihydrochloride), Mikromol
Specifications
Specifications
| Chemical Name or Material | Desethylhydroxychloroquine Dihydrochloride |
| CAS | 2732277-18-6 |
| Analyte or Component Names | Desethylhydroxychloroquine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H22 Cl N3 O . 2 Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Desethylhydroxychloroquine dihydrochloride, Cletoquine dihydrochloride, 2-[[(4RS)-4-[(7-Chloroquinolin-4-yl)amino]pentyl]amino]ethan-1-ol dihydrochloride, 2-[[4-[(7-Chloroquinolin-4-yl)amino]pentyl]amino]ethan-1-ol dihydrochloride, Hydroxychloroquine Sulfate Impurity C as Dihydrochloride, Hydroxychloroquine Impurity C as Dihydrochloride |
| InChI Formula | InChI=1S/C16H22ClN3O.2ClH/c1-12(3-2-7-18-9-10-21)20-15-6-8-19-16-11-13(17)4-5-14(15)16;;/h4-6,8,11-12,18,21H,2-3,7,9-10H2,1H3,(H,19,20);2*1H |
| Solution Type | Neat |
| SMILES | Cl.Cl.CC(CCCNCCO)Nc1ccnc2cc(Cl)ccc12 |
| Show More |