Learn More
Hydroxy Itraconazole-d8, MedChemExpress
Description
Hydroxy Itraconazole D8 is the deuterium labeled Hydroxy Itraconazole. Hydroxy Itraconazole is an active metabolite of Itraconazole (ITZ), which is a triazole antifungal agent.
Specifications
Specifications
| Chemical Name or Material | Hydroxy Itraconazole-d8 |
| Purity Grade Notes | Research |
| Physical Form | Powder |
| Molecular Formula | C35H30D8Cl2N8O5 |
| Color | White |
| Synonym | R-63373-d8 |
| SMILES | O=C1N(C(C([2H])([2H])[2H])([2H])C(O)([2H])C([2H])([2H])[2H])N=CN1C2=CC=C(N3CCN(C4=CC=C(OCC5OC(CN6N=CN=C6)(C7=CC=C(Cl)C=C7Cl)OC5)C=C4)CC3)C=C2 |
| Molecular Weight (g/mol) | 729.68 |
| Quantity | 1 mg |
| Formula Weight | 729.68 |
| Show More |