missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen peroxide solution, Honeywell™
Click to view available options
Quantity:
1 L
5 L
4 x 5 L
6 x 1 L
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
CAS Min % | ≥50.00%, ≥30.00% |
CAS Max % | <70.00%, <50.00% |
Packaging | Plastic Bottle |
Quantity | 4 x 5 L |
Physical Form | Liquid |
Grade | Reagent pharmacopeia chemistry reagents |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'