Learn More
Hydrogen peroxide 30%, BAKER ANALYZED™ ACS, J.T. Baker™
Product Code. p-7220851 Shop All J.T.Baker Products
Click to view available options
Quantity:
250 mL
2.5 L
Unit Size:
2.5 L
250mL
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15587984
Brand: JT Baker 2192.0250
Please to purchase this item. Need a web account? Register with us today!
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Exceeds ACS Specifications
  • Trace Impurities (in ppm):
    • Silver (Ag) max. 0.01
    • aluminum (Al) max. 0.05
    • Arsenic (As) max. 0.05
    • Barium (Ba) max. 0.02
    • Beryllium (Be) max. 0.01
    • Calcium (Ca) max. 0.5
    • Cadmium (Cd) max. 0.02
    • Cobalt (Co) max. 0.01
    • Chromium (Cr) max. 0.1
    • Copper (Cu) max. 0.01
    • Iron (Fe) max. 0.1
    • Germanium (Ge) max. 0.05
    • Potassium (K) max. 0.1
    • Lithium (Li) max. 0.01
    • Magnesium (Mg) max. 0.1
    • Manganese (Mn) max. 0.01
    • Molybdenum (Mo) max. 0.02
    • Sodium (Na) max. 0.5
    • Nickel (Ni) max. 0.05
    • Lead (Pb) max. 0.05
    • Strontium (Sr) max. 0.01
    • Titanium (Ti) max. 0.1
    • Thallium (Tl) max. 0.05
    • Vanadium (V) max. 0.01
    • Zinc (Zn) max. 0.05
    • Zirconium (Zr) max. 0.1
Chemical Identifiers
CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.01
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
IUPAC Name peroxol
SMILES OO
Specifications
Color Max. 10 (APHA)
Boiling Point ∼107°C (1013 hPa)
Melting Point ≤0°C
Assay Percent Range 30.0 to 32.0%
Density 1.11 g/cm³ (20°C)
Packaging Plastic Bottle
Residue after Evaporation max. 0.002%
Quantity 250 mL
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Oxidising liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : hydrogen peroxide