Learn More
Hydrocortisone 17-valerate, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Hydrocortisone Valerate |
| CAS | 57524-89-7 |
| Analyte or Component Names | Hydrocortisone Valerate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H38 O6 |
| Certifications/Compliance | ISO 17034 |
| Synonym | (11β)-11,21-Dihydroxy-17-[(1-oxopentyl)oxy]-pregn-4-ene-3,20-dione, Cortisol, 17-valerate (7CI), 11β,21-Dihydroxy-17α-valeryloxy-4-pregnene-3,20-dione, HydroVal, Hydrocortisone 17-valerate, Hydrocortisone pentanoate, Hydrocortisone valerate, Westcort |
| InChI Formula | InChI=1S/C26H38O6/c1-4-5-6-22(31)32-26(21(30)15-27)12-10-19-18-8-7-16-13-17(28)9-11-24(16,2)23(18)20(29)14-25(19,26)3/h13,18-20,23,27,29H,4-12,14-15H2,1-3H3/t18-,19-,20-,23+,24-,25-,26-/m0/s1 |
| Solution Type | Neat |
| SMILES | CCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3[C@@H](O)C[C@]12C)C(=O)CO |
| Show More |