missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TOR Antibody, R&D Systems™

Rat Monoclonal Antibody
196.00 € - 385.00 €
Specifications
| Antigen | TOR/mTOR |
|---|---|
| Clone | 986935 |
| Dilution | Immunohistochemistry 5-25μg/mL, Immunocytochemistry 8-25μg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18742328
![]() |
R&D Systems
MAB15371-SP |
25 μg |
196.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18743918
![]() |
R&D Systems
MAB15371-100 |
100 μg |
385.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TOR/mTOR Monoclonal specifically detects TOR/mTOR in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
| TOR/mTOR | |
| Immunohistochemistry 5-25μg/mL, Immunocytochemistry 8-25μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P42345 | |
| 2475 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 986935 | |
| Immunocytochemistry, Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| EC 2.7.11.1, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, FLJ44809, FRAP, FRAP1, FRAP1FKBP12-rapamycin complex-associated protein 1, FRAP2, Mammalian target of rapamycin, Mechanistic target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), mTOR, RAFT1, rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, Rapamycin target protein 1, RAPT1FKBP-rapamycin associated protein, serine/threonine-protein kinase mTOR | |
| E. coli-derived recombinant human TOR Phe1720-Ala2020 Accession # P42345 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human TOR in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title