missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TOR Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB15371-100
This item is not returnable.
View return policy
Description
TOR/mTOR Monoclonal specifically detects TOR/mTOR in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| TOR/mTOR | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| EC 2.7.11.1, FK506 binding protein 12-rapamycin associated protein 1, FK506 binding protein 12-rapamycin associated protein 2, FK506-binding protein 12-rapamycin complex-associated protein 1, FKBP12-rapamycin complex-associated protein, FLJ44809, FRAP, FRAP1, FRAP1FKBP12-rapamycin complex-associated protein 1, FRAP2, Mammalian target of rapamycin, Mechanistic target of rapamycin, mechanistic target of rapamycin (serine/threonine kinase), mTOR, RAFT1, rapamycin and FKBP12 target 1, rapamycin associated protein FRAP2, Rapamycin target protein 1, RAPT1FKBP-rapamycin associated protein, serine/threonine-protein kinase mTOR | |
| E. coli-derived recombinant human TOR Phe1720-Ala2020 Accession # P42345 | |
| 100 μg | |
| 2475 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunocytochemistry, Immunohistochemistry | |
| 986935 | |
| Immunohistochemistry 5-25μg/mL, Immunocytochemistry 8-25μg/mL | |
| P42345 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human TOR in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction