missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TFPI-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
188.00 € - 496.00 €
Specifications
| Antigen | TFPI-2 |
|---|---|
| Clone | 243227 |
| Dilution | Immunoprecipitation 25 ug/mL, Neutralization 0.45 ug/mL |
| Applications | Immunoprecipitation, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18734227
![]() |
R&D Systems
MAB25451-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18152471
![]() |
R&D Systems
MAB25451 |
500 μg |
496.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TFPI-2 Monoclonal specifically detects TFPI-2 in Human samples. It is validated for Immunoprecipitation, Neutralization.Specifications
| TFPI-2 | |
| Immunoprecipitation 25 ug/mL, Neutralization 0.45 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Placental protein 5, PP5, PP5FLJ21164, REF-1, TFPI2, TFPI-2REF1, tissue factor pathway inhibitor 2 | |
| TFPI2 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 243227 | |
| Immunoprecipitation, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human | |
| P48307 | |
| 7980 | |
| Mouse myeloma cell line NS0-derived recombinant human TFPI-2 Asp23-Phe235 Accession # P48307 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human TFPI-2 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title