missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human TFPI-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB25451
This item is not returnable.
View return policy
Description
TFPI-2 Monoclonal specifically detects TFPI-2 in Human samples. It is validated for Immunoprecipitation, Neutralization.
Specifications
| TFPI-2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Placental protein 5, PP5, PP5FLJ21164, REF-1, TFPI2, TFPI-2REF1, tissue factor pathway inhibitor 2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7980 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Immunoprecipitation, Neutralization | |
| 243227 | |
| Immunoprecipitation 25 ug/mL, Neutralization 0.45 ug/mL | |
| P48307 | |
| TFPI2 | |
| Mouse myeloma cell line NS0-derived recombinant human TFPI-2 Asp23-Phe235 Accession # P48307 | |
| 500 μg | |
| Primary | |
| Detects human TFPI-2 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction