missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P2X7/P2RX7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
208.00 € - 588.00 €
Specifications
| Antigen | P2X7/P2RX7 |
|---|---|
| Clone | 1058613 |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18785905
![]() |
R&D Systems
MAB97903-100 |
100 μg |
588.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18766205
![]() |
R&D Systems
MAB97903-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Human samples. It is validated for Flow CytometrySpecifications
| P2X7/P2RX7 | |
| Flow Cytometry 0.25 ug/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| ATP receptor, MGC20089, P2RX7, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A | |
| Human P2X7/P2RX7 synthetic peptide, Accession # Q99572 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1058613 | |
| Flow Cytometry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q99572 | |
| 5027 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title