Learn More
Human P2X7/P2RX7 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB97903-SP
 View more versions of this product
Product Code. 18766205
  • 208.00 € / 25µg
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
P2X7/P2RX7 Monoclonal antibody specifically detects P2X7/P2RX7 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
P2X7/P2RX7
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
ATP receptor, MGC20089, P2RX7, P2X purinoceptor 7, P2X7P2X7 receptor, P2X7R, P2Z receptor, Purinergic receptor, purinergic receptor P2X, ligand-gated ion channel, 7, purinergic receptor P2X7 variant A
Human P2X7/P2RX7 synthetic peptide, Accession # Q99572
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG2a
Flow Cytometry
1058613
Flow Cytometry 0.25 ug/10^6 cells
Q99572
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5027
Human
Purified