missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MUTED Partial ORF (NP_958437.1, 88 a.a. - 187 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_958437.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 63915 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139476
|
Abnova™
H00063915-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129476
|
Abnova™
H00063915-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a component of BLOC-1 (biogenesis of lysosome-related organelles complex 1). Components of this complex are involved in the biogenesis of organelles such as melanosomes and platelet-dense granules. A mouse model for Hermansky-Pudlak Syndrome is mutated in the murine version of this gene. Some transcripts of the downstream gene TXNDC5 overlap this gene, but they do not contain an open reading frame for this gene. [provided by RefSeq]
Sequence: EHTLPKCRDTMRDSLSQVLQRLQAANDSVCRLQQREQERKKIHSDHLVASEKQHMLQWDNFMKEQPNKRAEVDEEHRKAMERLKEQYAEMEKDLAKFSTFSpecifications
NP_958437.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E2287/MU | |
MUTED | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
63915 | |
MUTED (Human) Recombinant Protein (Q01) | |
EHTLPKCRDTMRDSLSQVLQRLQAANDSVCRLQQREQERKKIHSDHLVASEKQHMLQWDNFMKEQPNKRAEVDEEHRKAMERLKEQYAEMEKDLAKFSTF | |
RUO | |
MUTED | |
Wheat Germ (in vitro) | |
GST | |
Liquid |