Learn More
Abnova™ Human MUTED Partial ORF (NP_958437.1, 88 a.a. - 187 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00063915-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a component of BLOC-1 (biogenesis of lysosome-related organelles complex 1). Components of this complex are involved in the biogenesis of organelles such as melanosomes and platelet-dense granules. A mouse model for Hermansky-Pudlak Syndrome is mutated in the murine version of this gene. Some transcripts of the downstream gene TXNDC5 overlap this gene, but they do not contain an open reading frame for this gene. [provided by RefSeq]
Sequence: EHTLPKCRDTMRDSLSQVLQRLQAANDSVCRLQQREQERKKIHSDHLVASEKQHMLQWDNFMKEQPNKRAEVDEEHRKAMERLKEQYAEMEKDLAKFSTFSpecifications
NP_958437.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
EHTLPKCRDTMRDSLSQVLQRLQAANDSVCRLQQREQERKKIHSDHLVASEKQHMLQWDNFMKEQPNKRAEVDEEHRKAMERLKEQYAEMEKDLAKFSTF | |
RUO | |
MUTED | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
63915 | |
MUTED (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686E2287/MU | |
MUTED | |
Recombinant | |
wheat germ expression system |