Learn More
Abnova™ Human MPP4 Partial ORF (NP_149055, 1 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00058538-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) protein family, with an N-terminal PDZ domain, a central src homology 3 region (SH3), and a C-terminal guanylate kinase-like (GUK) domain. The protein is localized to the outer limiting membrane in the retina, and is thought to function in photoreceptor polarity and the organization of specialized intercellular junctions. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]
Sequence: MIQSDKGADPPDKKDMKLSTATNPQNGLSQILRLVLQELSLFYSRDVNGVCLLYDLLHSPWLQALLKIYDCLQEFKEKKLVPATPHAQVLSYEVVELLRETPTSPEIQESpecifications
NP_149055 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MIQSDKGADPPDKKDMKLSTATNPQNGLSQILRLVLQELSLFYSRDVNGVCLLYDLLHSPWLQALLKIYDCLQEFKEKKLVPATPHAQVLSYEVVELLRETPTSPEIQE | |
RUO | |
MPP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
58538 | |
MPP4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALS2CR5/DLG6 | |
MPP4 | |
Recombinant | |
wheat germ expression system |