missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human MPP4 Partial ORF (NP_149055, 1 a.a. - 109 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00 € - 508.00 €
Specifications
Accession Number | NP_149055 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 58538 |
Molecular Weight (g/mol) | 37.73kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16139346
|
Abnova™
H00058538-Q01.25UG |
25 ug |
508.00 €
25µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16129346
|
Abnova™
H00058538-Q01.10UG |
10 ug |
335.00 €
10µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the membrane-associated guanylate kinase (MAGUK) protein family, with an N-terminal PDZ domain, a central src homology 3 region (SH3), and a C-terminal guanylate kinase-like (GUK) domain. The protein is localized to the outer limiting membrane in the retina, and is thought to function in photoreceptor polarity and the organization of specialized intercellular junctions. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]
Sequence: MIQSDKGADPPDKKDMKLSTATNPQNGLSQILRLVLQELSLFYSRDVNGVCLLYDLLHSPWLQALLKIYDCLQEFKEKKLVPATPHAQVLSYEVVELLRETPTSPEIQESpecifications
NP_149055 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ALS2CR5/DLG6 | |
MPP4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
58538 | |
MPP4 (Human) Recombinant Protein (Q01) | |
MIQSDKGADPPDKKDMKLSTATNPQNGLSQILRLVLQELSLFYSRDVNGVCLLYDLLHSPWLQALLKIYDCLQEFKEKKLVPATPHAQVLSYEVVELLRETPTSPEIQE | |
RUO | |
MPP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |